MilliporeSigma
  • Novel naphtho[2,1-d]oxazole-4,5-diones as NQO1 substrates with improved aqueous solubility: Design, synthesis, and in vivo antitumor evaluation.
Novel naphtho[2,1-d]oxazole-4,5-diones as NQO1 substrates with improved aqueous solubility: Design, synthesis, and in vivo antitumor evaluation.
Bioorganic & medicinal chemistry (2016-01-25)
Xiang Li, Jinlei Bian, Nan Wang, Xue Qian, Jing Gu, Tong Mu, Jun Fan, Xiuwen Yang, Shangzhen Li, Tingting Yang, Haopeng Sun, Qidong You, Xiaojin Zhang
ABSTRACT
A new series of ortho-naphthoquinone analogs of β-lapachone were designed, synthesized and evaluated. The biological results indicated that most of our compounds were efficient substrates for NQO1. The new scaffold with water-soluble side chain resulted in greater solubility under acidic condition compared to β-lapachone. Thus avoiding the use of hydroxylpropyl β-cyclodextrin which would finally cause the rapid drug clearance from the blood and dose-limiting toxicity in the form of hemolytic anemia. The most soluble and promising compound in this series was 2-((4-benzylpiperazin-1-yl)methyl)naphtho[2,1-d]oxazole-4,5-dione (3k), which inhibited cancer cell (NQO1-rich A549 cell line) growth at IC50 values of 4.6±1.0μmol·L(-1). Furthermore, compound 3k had in vivo antitumor activity in an A549 tumor xenografts mouse model comparable to the activity obtained with β-lapachone. The results indicated that these ortho-naphthoquinones could serve as promising leads for further optimization as novel substrates for NQO1.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
NQO1 human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)