MilliporeSigma
  • Optimized heart rate for 320-row cardiac CT can be feasibly predicted from prescan parameters.
Optimized heart rate for 320-row cardiac CT can be feasibly predicted from prescan parameters.
SpringerPlus (2015-11-21)
Eriko Maeda, Nobuo Tomizawa, Kodai Yamamoto, Shigeaki Kanno, Masaaki Akahane, Kenji Ino, Masae Uehara, Aiko Sakamoto, Toshiaki Semboku, Rumiko Torigoe, Kuni Ohtomo
ABSTRACT
To evaluate the degree of heart rate (HR) changes at rest (HRrest), during breath hold (HRtest), and during cardiac CT examinations (HRscan) in a large group of patients , and to derive and asses the feasibility of a predictive formula for HRscan. HRrest, HRtest, and HRscan were retrospectively compared in a total of 563 consecutive patients who underwent 320-row cardiac CT. Multiple regression analysis was performed to derive predictive formulae for HRscan in the entire study population and, in each group of patients with decreased (Dec) or increased (Inc) HR during breath hold. The predictive formula was evaluated as accurate when less than 5 % of the actual HRscan exceeded the predicted HRscan by ±5 beats per minute (bpm). The average values of the HRtest (65.3 ± 12.0 bpm) and HRscan (63.7 ± 11.9 bpm) significantly decreased from those of the HRrest (68.4 ± 11.9 bpm) (p < 0.0001). The predictive formula (HRscan = 3.601 + 0.113HRrest + 0.8HRtest) was determined to be accurate only in Group Dec. The HRtest significantly decreased from the HRrest, and the HRscan significantly decreased from the HRtest. An accurate predictive formula for HRscan could be built only for Group Dec.
Product Description
Isosorbide dinitrate, European Pharmacopoeia (EP) Reference Standard