MilliporeSigma
  • Stimulating effects of a diet negative in dietary cation-anion difference on calcium absorption from the rumen in sheep.
Stimulating effects of a diet negative in dietary cation-anion difference on calcium absorption from the rumen in sheep.
Journal of animal physiology and animal nutrition (2015-02-04)
M R Wilkens, C Praechter, G Breves, B Schröder
ABSTRACT
The concept of feeding anionic salts in late gestation is widely used to prevent milk fever in dairy cows. While the effects of these diets on renal Ca excretion and tissue responsiveness towards parathyroid hormone have clearly been demonstrated, data on a potential impact on gastrointestinal Ca absorption are conflicting. Therefore, the aim of this study was to investigate the influence of feeding a diet negative in dietary cation-anion difference (DCAD) on ruminal mineral concentrations, fermentation products, electrophysiological properties of rumen epithelia and Ca flux rates. For this purpose, sheep were kept for 3 weeks on diets that were either positive or negative in DCAD. The induction of a compensated hyperchloremic metabolic acidosis could be demonstrated by increased plasma Cl and enhanced concentrations of ionised Ca, while plasma concentrations of HCO3- and base excess were decreased with the low DCAD diet. Neither transmural potential differences nor fermentation products were affected, but ruminal concentrations of Cl and Mg as well as the relation of ionised to total Ca were increased. Ussing chamber experiments revealed alterations of electrophysiological parameters and an increase in the electroneutral component of Ca flux rates from the mucosal to the serosal side of rumen epithelium. As plasma calcitriol concentrations were not affected, it can be concluded that the administration of anionic salts results in a vitamin D-independent stimulation of ruminal Ca transport.
Sigma-Aldrich
Di-(4-chlorobenzyl)azodicarboxylate, 97%