MilliporeSigma
"Size-Independent" Single-Electron Tunneling.
The journal of physical chemistry letters (2015-12-01)
Jianli Zhao, Shasha Sun, Logan Swartz, Shawn Riechers, Peiguang Hu, Shaowei Chen, Jie Zheng, Gang-Yu Liu
ABSTRACT
Incorporating single-electron tunneling (SET) of metallic nanoparticles (NPs) into modern electronic devices offers great promise to enable new properties; however, it is technically very challenging due to the necessity to integrate ultrasmall (<10 nm) particles into the devices. The nanosize requirements are intrinsic for NPs to exhibit quantum or SET behaviors, for example, 10 nm or smaller, at room temperature. This work represents the first observation of SET that defies the well-known size restriction. Using polycrystalline Au NPs synthesized via our newly developed solid-state glycine matrices method, a Coulomb Blockade was observed for particles as large as tens of nanometers, and the blockade voltage exhibited little dependence on the size of the NPs. These observations are counterintuitive at first glance. Further investigations reveal that each observed SET arises from the ultrasmall single crystalline grain(s) within the polycrystal NP, which is (are) sufficiently isolated from the nearest neighbor grains. This work demonstrates the concept and feasibility to overcome orthodox spatial confinement requirements to achieve quantum effects.
Sigma-Aldrich
1-Octanethiol, ≥98.5%