MilliporeSigma
  • High-performance liquid chromatographic separations of stereoisomers of chiral basic agrochemicals with polysaccharide-based chiral columns and polar organic mobile phases.
High-performance liquid chromatographic separations of stereoisomers of chiral basic agrochemicals with polysaccharide-based chiral columns and polar organic mobile phases.
Journal of separation science (2015-10-27)
Iza Matarashvili, Iamze Shvangiradze, Lali Chankvetadze, Shota Sidamonidze, Nino Takaishvili, Tivadar Farkas, Bezhan Chankvetadze
ABSTRACT
The separation of the stereoisomers of 23 chiral basic agrochemicals was studied on six different polysaccharide-based chiral columns in high-performance liquid chromatography with various polar organic mobile phases. Along with the successful separation of analyte stereoisomers, emphasis was placed on the effect of the chiral selector and mobile phase composition on the elution order of stereoisomers. The interesting phenomenon of reversal of enantiomer/stereoisomer elution order function of the polysaccharide backbone (cellulose or amylose), type of derivative (carbamate or benzoate), nature, and position of the substituent(s) in the phenylcarbamate moiety (methyl or chloro) and the nature of the mobile phase was observed. For several of the analytes containing two chiral centers all four stereoisomers were resolved with at least one chiral selector/mobile phase combination.
Product Description
Supelco
Prothioconazole-desthio, PESTANAL®, analytical standard
Supelco
Metconazole, PESTANAL®, analytical standard, mixture of stereoisomers
Supelco
Prothioconazole, PESTANAL®, analytical standard
Supelco
Triticonazole, PESTANAL®, analytical standard