MilliporeSigma
  • Inflammatory regulation of steroid sulfatase: A novel mechanism to control estrogen homeostasis and inflammation in chronic liver disease.
Inflammatory regulation of steroid sulfatase: A novel mechanism to control estrogen homeostasis and inflammation in chronic liver disease.
Journal of hepatology (2015-07-30)
Mengxi Jiang, Marcus Klein, Ulrich M Zanger, Mohammad K Mohammad, Matthew C Cave, Nilesh W Gaikwad, Natasha J Dias, Kyle W Selcer, Yan Guo, Jinhan He, Xiuhui Zhang, Qiujin Shen, Wenxin Qin, Jiang Li, Song Li, Wen Xie
ABSTRACT
Chronic inflammatory liver diseases are associated with estrogen excess and feminization in men, which is thought to be due to compromised liver function to break down estrogens. The goal of this study is to determine whether the inflammatory induction of steroid sulfatase (STS), which converts inactive estrogen sulfates to active estrogens, may have contributed to the estrogen excess in chronic liver disease. We performed bioinformatic analysis, real-time PCR, immunohistochemistry, and UPLC/MS-MS to analyze hepatic STS expression and serum estrogen levels in patients with chronic liver diseases. The crosstalk between NF-κB pathway and STS-regulated estrogen signaling was investigated by electrophoretic mobility shift assay, chromatin immunoprecipitation, luciferase assay and gene knockdown experiments in human hepatocytes. Hepatic STS was induced in patients with chronic inflammatory liver diseases, which was accompanied by increased circulating estrogen levels. The human STS gene, but not the mouse Sts gene, was induced by inflammatory stimuli in hepatic cells. Mechanistically, STS was established as a novel NF-κB target gene, whose induction facilitated the conversion of inactive estrogen sulfates to active estrogens, and consequently attenuated the inflammatory response. In contrast, genetic or pharmacological inhibition of STS or a direct blockade of estrogen signaling sensitized liver cells to the transcriptional activation of NF-κB and inflammatory response, possibly through the inhibition of IκB kinase activation. Our results suggest a negative feedback loop in chronic inflammatory liver diseases, in which the inflammatory activation of NF-κB induces STS gene expression. The induced STS facilitates the conversion of inactive estrogen sulfates to active estrogens, which in return attenuates the NF-κB-mediated inflammation.
Sigma-Aldrich
MISSION® esiRNA, targeting human STS
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
STX64, ≥98% (HPLC)
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Sigma-Aldrich
Poly(deoxyinosinic-deoxycytidylic) acid sodium salt, double-stranded alternating copolymer