MilliporeSigma
  • Human chorionic gonadotropin-dependent regulation of 17beta-hydroxysteroid dehydrogenase type 4 in preovulatory follicles and its potential role in follicular luteinization.
Human chorionic gonadotropin-dependent regulation of 17beta-hydroxysteroid dehydrogenase type 4 in preovulatory follicles and its potential role in follicular luteinization.
Endocrinology (2004-01-17)
Kristy A Brown, Derek Boerboom, Nadine Bouchard, Monique Doré, Jacques G Lussier, Jean Sirois
ABSTRACT
17Beta-hydroxysteroid dehydrogenase type 4 (17betaHSD4) has a unique multidomain structure, with one domain involved in 17beta-estradiol inactivation. The objective of the study was to investigate the regulation of 17betaHSD4 during human chorionic gonadotropin (hCG)-induced ovulation/luteinization. The equine 17betaHSD4 cDNA was cloned and was shown to encode a 735-amino acid protein that is highly conserved (81-87% identity) compared with other mammalian orthologs. RT-PCR/Southern blot analyses were performed to study the regulation of 17betaHSD4 transcripts in equine preovulatory follicles isolated between 0-39 h after hCG treatment. Results showed the presence of basal 17betaHSD4 mRNA expression before hCG treatment, but an increase was observed in follicles obtained 24 h after hCG (P < 0.05). Analyses of isolated preparations of granulosa and theca interna cells identified basal mRNA expression in both layers, but granulosa cells appeared as the predominant site of follicular 17betaHSD4 mRNA induction. A specific polyclonal antibody was raised against a fragment of the equine protein and used to study regulation of the 17betaHSD4 protein. Immunoblots showed an increase in full-length 17betaHSD4 protein in follicles 24 h after hCG (P < 0.05), in keeping with mRNA results. Immunohistochemical data confirmed the induction of the enzyme in follicular cells after hCG treatment. Collectively, these results demonstrate that the gonadotropin-dependent induction of follicular luteinization is accompanied by an increase in 17betaHSD4 expression. Considering the estrogen-inactivating function of 17betaHSD4, its regulated expression in luteinizing preovulatory follicles appears as a potential complementary mechanism to reduce circulating levels of 17beta-estradiol after the LH surge.