MilliporeSigma
  • High prevalence of the GSTM3*A/B polymorphism in sub-Sarahan African populations.
High prevalence of the GSTM3*A/B polymorphism in sub-Sarahan African populations.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas (2010-06-16)
D Teixeira, D Vargens, A Príncipe, E Oliveira, A Amorim, M J Prata, G Suarez-Kurtz
ABSTRACT
A 3-bp insertion/deletion polymorphism in intron 6 of GSTM3 (rs1799735, GSTM3*A/*B) affects the activity of the phase 2 xenobiotic metabolizing enzyme GSTM3 and has been associated with increased cancer risk. The GSTM3*B allele is rare or absent in Southeast Asians, occurs in 5-20% of Europeans but was detected in 80% of Bantu from South Africa. The wide genetic diversity among Africans led us to investigate whether the high frequency of GSTM3*B prevailed in other sub-Saharan African populations. In 168 healthy individuals from Angola, Mozambique and the São Tomé e Príncipe islands, the GSTM3*B allele was three times more frequent (0.74-0.78) than the GSTM3*A allele (0.22-0.26), with no significant differences in allele frequency across the three groups. We combined these data with previously published results to carry out a multidimensional scaling analysis, which provided a visualization of the worldwide population affinities based on the GSTM3 *A/*B polymorphism.
Sigma-Aldrich
GST M3-3, Recombinant Human