MilliporeSigma
  • Gas phase thermal reactions of exo-8-cyclopropyl-bicyclo[4.2.0]oct-2-ene (1-exo).
Gas phase thermal reactions of exo-8-cyclopropyl-bicyclo[4.2.0]oct-2-ene (1-exo).
Molecules (Basel, Switzerland) (2014-01-30)
Phyllis A Leber, Anthony J Nocket, William Hancock-Cerutti, Christopher Y Bemis, Wint Khant Khine, Joseph A Mohrbacher Iii, John E Baldwin
ABSTRACT
The title compound 1-exo (with minor amounts of its C8 epimer 1-endo) was prepared by Wolff-Kishner reduction of the cycloadduct of 1,3-cyclohexadiene and cyclopropylketene. The [1,3]-migration product 2-endo was synthesized by efficient selective cyclopropanation of endo-5-vinylbicyclo[2.2.2]oct-2-ene at the exocyclic π-bond. Gas phase thermal reactions of 1-exo afforded C8 epimerization to 1-endo, [1,3]- migrations to 2-exo and 2-endo, direct fragmentation to cyclohexadiene and vinylcyclopropane, and CPC rearrangement in the following relative kinetic order: kep > k13 > kf > kCPC.
Sigma-Aldrich
Cyclopropylacetonitrile, 97%