MilliporeSigma
  • A Mild, Large-Scale Synthesis of 1,3-Cyclooctanedione: Expanding Access to Difluorinated Cyclooctyne for Copper-Free Click Chemistry.
A Mild, Large-Scale Synthesis of 1,3-Cyclooctanedione: Expanding Access to Difluorinated Cyclooctyne for Copper-Free Click Chemistry.
Tetrahedron letters (2011-06-29)
Evan A Sims, Cole A Deforest, Kristi S Anseth
ABSTRACT
We report the large-scale synthesis of 1,3-cyclooctanedione in five steps with 29% yield. This molecule is a synthetic precurser to difluorinated cyclooctyne, which participates in a bioorthogonal copper-free click reaction with azides. The final step demonstrates the first successful application of the Wacker-Tsuji oxidation to form a cyclic 1,3-dione.