MilliporeSigma
  • Counterintuitive compaction behavior of clopidogrel bisulfate polymorphs.
Counterintuitive compaction behavior of clopidogrel bisulfate polymorphs.
Journal of pharmaceutical sciences (2012-04-11)
Kailas S Khomane, Parth K More, Arvind K Bansal
ABSTRACT
Being a density violator, clopidogrel bisulfate (CLP) polymorphic system (forms I and II) allows us to study individually the impact of molecular packing (true density) and thermodynamic properties such as heat of fusion on the compaction behavior. These two polymorphs of CLP were investigated for in-die and out-of-die compaction behavior using CTC profile, Heckel, and Walker equations. Compaction studies were performed on a fully instrumented rotary tabletting machine. Detailed examinations of the molecular packing of each form revealed that arrangement of the sulfate anion differs significantly in both crystal forms, thus conferring different compaction behavior to two forms. Close cluster packing of molecules in form I offers a rigid structure, which has poor compressibility and hence resists deformation under compaction pressure. This results into lower densification, higher yield strength, and mean yield pressure, as compared with form II at a given pressure. However, by virtue of higher bonding strength, form I showed superior tabletability, despite its poor compressibility and deformation behavior. Form I, having higher true density and lower heat of fusion showed higher bonding strength. Hence, true density and not heat of fusion can be considered predictor of bonding strength of the pharmaceutical powders.
Product Description
USP
Clopidogrel bisulfate, United States Pharmacopeia (USP) Reference Standard