MilliporeSigma
  • Sterically non-hindering endocyclic ligands of the bi-isoquinoline family.
Sterically non-hindering endocyclic ligands of the bi-isoquinoline family.
Chemical communications (Cambridge, England) (2005-12-24)
Fabien Durola, Jean-Pierre Sauvage, Oliver S Wenger
ABSTRACT
Bi-isoquinoline can be used as a building block to prepare a new family of non-sterically hindering chelates, including a macrocyclic system; the endocyclic nature of the ligands has been confirmed by the X-ray structure of an octahedral tris-chelate iron(II) complex, which shows that the three chelates are easily accommodated in the coordination sphere of the metal in spite of their crescent shape.