MilliporeSigma
  • Seasonal effects of the UCP3 and the RPTOR gene polymorphisms on obesity traits in Japanese adults.
Seasonal effects of the UCP3 and the RPTOR gene polymorphisms on obesity traits in Japanese adults.
Journal of physiological anthropology (2014-12-24)
Kazuhiro Nakayama, Hiroshi Miyashita, Sadahiko Iwamoto
ABSTRACT
Non-shivering thermogenesis (NST) involves a substantial amount of energy expenditure in humans and, thus, contributes to reducing the risk for obesity. Molecular evolutionary studies have reported that SNPs in/near the uncoupling protein 3 gene (UCP3) and the regulatory associated protein of mTOR complex 1 gene (RPTOR) might influence NST and confer adaptive advantages for modern human dispersal into cold environments. In the present study, the impact of these SNPs on obesity-related traits was investigated. Study subjects consisted of 2,834 Japanese adults (percentage of female: 46%, mean age: 51.5). Associations of the UCP3-55C/T and the RPTOR-26934C/T - the 2 potential genetic variations involved in cold adaptation and thermogenic mechanisms in mammals, with quantitative obesity-related traits including body mass index (BMI), waist circumference, visceral fat area (VFA), VFA adjusted for BMI, and selected blood parameters - were tested using multiple linear regression models. Sliding windowsampling analysis was applied to depict seasonal effects of the SNPs on the obesity-related phenotypes. UCP3-55C/T and the RPTOR-26934C/T did not show any association with obesity traits and blood chemical parameters in multiple linear regression models consisting of the whole subjects. Moreover, sliding window sampling-based association analyses involving seasonality also failed to find associations between these two SNPs and obesity-related traits. UCP3-55C/T and the RPTOR-26934C/T may only have subtle effects on the development of obesity-related traits in the present humans. These two SNPs might be irrelevant to inter-individual variations in energy metabolism and efficiency of NST.
Sigma-Aldrich
mTOR/Raptor/MLST8 human, recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)