MilliporeSigma
  • ITIH3 polymorphism may confer susceptibility to psychiatric disorders by altering the expression levels of GLT8D1.
ITIH3 polymorphism may confer susceptibility to psychiatric disorders by altering the expression levels of GLT8D1.
Journal of psychiatric research (2014-01-01)
Daimei Sasayama, Hiroaki Hori, Noriko Yamamoto, Seiji Nakamura, Toshiya Teraishi, Masahiko Tatsumi, Kotaro Hattori, Miho Ota, Teruhiko Higuchi, Hiroshi Kunugi
ABSTRACT
A recent genome-wide analysis indicated that a polymorphism (rs2535629) of ITIH3 showed the strongest association signal with susceptibility to psychiatric disorders in Caucasian populations. The aim of the study was to replicate the association of rs2535629 with schizophrenia and major depressive disorder (MDD) in Japanese subjects. A total of 611 patients with schizophrenia, 868 with MDD, and 1193 healthy controls were successfully genotyped for rs2535629. A significant difference in allele distribution was found between patients with schizophrenia and controls (odds ratio [OR] = 1.21, 95% confidence interval [CI]: 1.05-1.39, P = 0.0077). A similar trend was found for patients with MDD (OR = 1.11, 95% CI: 0.98-1.26, P = 0.092). The allele distribution in the combined patient group (schizophrenia and MDD) was significantly different from that of the control group (OR = 1.15, 95% CI: 1.03-1.28, P = 0.011). Gene expression microarray analysis of whole blood samples in 39 MDD patients and 40 healthy controls showed that rs2535629 has a strong influence on the expression levels of ITIH4 and GLT8D1. The expression levels of GLT8D1 were significantly higher in patients with MDD than in controls (P = 0.021). To our knowledge, the present study showed for the first time the association of rs2535629 with psychiatric disorders in an Asian population. Our findings suggest that rs2535629 influences the susceptibility to psychiatric disorders by affecting the expression level of GLT8D1.