MilliporeSigma
  • Cisplatin-induced acute renal failure is associated with an increase in the cytokines interleukin (IL)-1beta, IL-18, IL-6, and neutrophil infiltration in the kidney.
Cisplatin-induced acute renal failure is associated with an increase in the cytokines interleukin (IL)-1beta, IL-18, IL-6, and neutrophil infiltration in the kidney.
The Journal of pharmacology and experimental therapeutics (2007-04-03)
Sarah Faubel, Eli C Lewis, Leonid Reznikov, Danica Ljubanovic, Thomas S Hoke, Hilary Somerset, Dong-Jin Oh, Lawrence Lu, Christina L Klein, Charles A Dinarello, Charles L Edelstein
ABSTRACT
We have demonstrated that caspase-1-deficient (caspase-1(-/-)) mice are functionally and histologically protected against cisplatin-induced acute renal failure (ARF). Caspase-1 exerts proinflammatory effects via the cytokines interleukin (IL)-1beta, IL-18, IL-6, and neutrophil recruitment. We sought to determine the role of the cytokines IL-1beta, IL-18, and IL-6 and neutrophil recruitment in cisplatin-induced ARF. We first examined IL-1beta; renal IL-1beta increased nearly 2-fold in cisplatin-induced ARF and was reduced in the caspase-1(-/-) mice. However, inhibition with IL-1 receptor antagonist (IL-1Ra) did not attenuate cisplatin-induced ARF. Renal IL-18 increased 2.5-fold; however, methods to inhibit IL-18 using IL-18 antiserum and transgenic mice that overproduce IL-18-binding protein (a natural inhibitor of IL-18) did not protect. Renal IL-6 increased 3-fold; however, IL-6-deficient (IL-6(-/-)) mice still developed cisplatin-induced ARF. We next examined neutrophils; blood neutrophils increased dramatically after cisplatin injection; however, prevention of peripheral neutrophilia and renal neutrophil infiltration with the neutrophil-depleting antibody RB6-8C5 did not protect against cisplatin-induced ARF. In summary, our data demonstrated that cisplatin-induced ARF is associated with increases in the cytokines IL-1beta, IL-18, and IL-6 and neutrophil infiltration in the kidney. However, inhibition of IL-1beta, IL-18, and IL-6 or neutrophil infiltration in the kidney is not sufficient to prevent cisplatin-induced ARF.
Sigma-Aldrich
REDExtract-N-Amp Tissue PCR Kit, sufficient for 10 reactions, sufficient for 100 reactions, sufficient for 1000 reactions, hotstart, dNTPs included