MilliporeSigma
  • Involvement of genetic factors in the response to a variable-dosing ranibizumab treatment regimen for age-related macular degeneration.
Involvement of genetic factors in the response to a variable-dosing ranibizumab treatment regimen for age-related macular degeneration.
Molecular vision (2010-12-15)
Slawomir J Teper, Anna Nowinska, Jaroslaw Pilat, Andrzej Palucha, Edward Wylegala
ABSTRACT
To determine whether gene polymorphisms of the major genetic risk factor for age-related macular susceptibility 2 (ARMS2 A69S) and the complement factor H Y402H influence the response to a variable-dosing treatment regimen with ranibizumab for age-related macular degeneration. This prospective cohort study included 90 patients (90 eyes) with exudative age related macular degeneration (AMD) treated with ranibizumab. Patients underwent a 1-year treatment as in the Study of Ranibizumab in Patients with Subfoveal Choroidal Neovascularization Secondary to Age-Related Macular Degeneration (Mitchell et al.). Injections were administered monthly when a patient lost five letters on the Early Treatment Diabetic Retinopathy Study chart or gained 100 μm in central subfield retinal thickness (CSRT). Genotypes (rs10490924 and rs1061170) were analyzed using gene sequence analysis. Best-corrected visual acuity (BCVA) and CSRT values were compared between ARMS2 and complement factor H genotypes. Multiple regression analysis was used to assess the statistical significance. Mean increase in visual acuity was 4.44±8.12 letters with a 103.63±94.7 µm decrease in CSRT. BCVA improvement was statistically significant in all genotype groups except in homozygous 69S in the AMRS2 gene. CSRT and BCVA changes were correlated (r=0.2521; 95% CI: 0.04746-0.4364, p=0.0165). Multiple regression analysis revealed a significant impact of 69S (p=0.015) on the change in BCVA. Visual acuity did not improve during the study in patients homozygous for ARMS2 69S, despite a decrease in CSRT. Further investigation is needed to confirm our findings and understand the mechanisms involved.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 10 extractions, sufficient for 10 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 100 extractions, sufficient for 500 amplifications
Pricing and availability is not currently available.