MilliporeSigma
  • Anti-AIDS agents 79. Design, synthesis, molecular modeling and structure-activity relationships of novel dicamphanoyl-2',2'-dimethyldihydropyranochromone (DCP) analogs as potent anti-HIV agents.
Anti-AIDS agents 79. Design, synthesis, molecular modeling and structure-activity relationships of novel dicamphanoyl-2',2'-dimethyldihydropyranochromone (DCP) analogs as potent anti-HIV agents.
Bioorganic & medicinal chemistry (2010-08-24)
Ting Zhou, Qian Shi, Chin-Ho Chen, Hao Zhu, Li Huang, Phong Ho, Kuo-Hsiung Lee
ABSTRACT
In a continued study, 23 3'R,4'R-di-O-(-)-camphanoyl-2',2'-dimethyldihydropyrano[2,3-f]chromone (DCP) derivatives (5-27) were synthesized, and screened for anti-HIV activity against both a non-drug-resistant NL4-3 strain and multiple reverse transcriptase (RT) inhibitor-resistant (RTMDR-1) strain, using 2-EDCP (4) and 2-MDCP (35) as controls. New DCP analogs 5, 9, 14, and 22 exhibited potent anti-HIV activity against HIVNL4-3 with EC50 and therapeutic index (TI) values ranging from 0.036 microM to 0.14 microM and from 110 to 420, respectively. Compounds 5 and 9 also exhibited good activity against RTMDR-1 (EC50 0.049 and 0.054 microM; TI 310 and 200, respectively), and were twofold more potent than the leads 4 and 35 (EC50 0.11 and 0.19 microM; TI 60 and 58, respectively). Evaluation of water solubility showed that 5 and 22 were 5-10 times more water soluble than 4. Quantitative structure-activity relationship (QSAR) modeling results were first performed on this compound type, and the models should aid in design of future anti-HIV DCP analogs and potential clinical drug candidates.