MilliporeSigma
An efficient Agrobacterium-mediated transformation system for poplar.
International journal of molecular sciences (2014-06-17)
Ali Movahedi, Jiaxin Zhang, Rasoul Amirian, Qiang Zhuge
ABSTRACT
Poplar is a model system for the regeneration and genetic transformation of woody plants. To shorten the time required for studies of transgenic poplar, efforts have been made to optimize transformation methods that use Agrobacterium tumefaciens. In this study, an Agrobacterium infective suspension was treated at 4 °C for at least 10 h before infecting explants. By transforming the Populus hybrid clone "Nanlin895" (Populus deltoides×P. euramericana) with Agrobacterium harboring the PBI121:CarNAC6 binary vector, we showed that the transformation efficiency was improved significantly by multiple independent factors, including an Agrobacterium infective suspension with an OD600 of 0.7, an Agrobacterium infection for 120 min, an Agrobacterium infective suspension at a pH of 5.0, an acetosyringone concentration of 200 µM, a cocultivation at 28 °C, a cocultivation for 72 h and a sucrose concentration of 30 g/L in the cocultivation medium. We also showed that preculture of wounded leaf explants for two days increased the regeneration rate. The integration of the desired gene into transgenic poplars was detected using selective medium containing kanamycin, followed by southern blot analysis. The expression of the transgene in the transgenic lines was confirmed by northern blot analysis.
Product Description
Roche
DIG-High Prime DNA Labeling and Detection Starter Kit I, sufficient for 12 labeling reactions, sufficient for 24 blots, suitable for hybridization, suitable for Northern blotting, suitable for Southern blotting