MilliporeSigma
  • Di-(2-ethylhexyl) phthalate inhibits DNA replication leading to hyperPARylation, SIRT1 attenuation, and mitochondrial dysfunction in the testis.
Di-(2-ethylhexyl) phthalate inhibits DNA replication leading to hyperPARylation, SIRT1 attenuation, and mitochondrial dysfunction in the testis.
Scientific reports (2014-09-23)
Xiaolin Li, Evandro Fei Fang, Morten Scheibye-Knudsen, Honghua Cui, Lu Qiu, Jian Li, Yuping He, Jing Huang, Vilhelm A Bohr, Tzi Bun Ng, Hongwei Guo
ABSTRACT
Di-(2-ethylhexyl)-phthalate (DEHP) is a ubiquitously used endocrine disruptor.There is widespread exposure to DEHP in the general population which has raised substantial public concern due to its potential detrimental health effects. It is particularly pertinent to investigate the molecular mechanisms of its testicular toxicity which are largely unknown. By feeding male rats DEHP for 2 weeks, rat spermatogenesis became disrupted, resulting in a decreased number of spermatocytes and spermatids. Since rapidly dividing tissues appeared to be particularly vulnerable to DEHP toxicity we investigated the effect of DEHP on DNA replication. Intriguingly, DEHP appeared to inhibit DNA replication as evidenced by results of fiber tract analysis. This led to induction of the mitochondrial apoptotic pathways and increased ROS production. Furthermore, the toxicity of DEHP led to respiratory chain defects and attenuation of ATP level probably brought about by hyperPARylation and undermined SIRT1 activity. Our findings reveal a previously unknown mitochondrial dysfunction in DEHP-induced testicular toxicity and highlight the importance of SIRT1 in male reproduction.
Product Description
Roche
In Situ Cell Death Detection Kit, AP, sufficient for ≤50 tests, kit of 1 (3 components), suitable for detection