MilliporeSigma
  • Among developmental regulators, StuA but not BrlA is essential for penicillin V production in Penicillium chrysogenum.
Among developmental regulators, StuA but not BrlA is essential for penicillin V production in Penicillium chrysogenum.
Applied and environmental microbiology (2010-12-15)
Claudia Sigl, Hubertus Haas, Thomas Specht, Kristian Pfaller, Hubert Kürnsteiner, Ivo Zadra
ABSTRACT
In filamentous fungi, secondary metabolism is often linked with developmental processes such as conidiation. In this study we analyzed the link between secondary metabolism and conidiation in the main industrial producer of the β-lactam antibiotic penicillin, the ascomycete Penicillium chrysogenum. Therefore, we generated mutants defective in two central regulators of conidiation, the transcription factors BrlA and StuA. Inactivation of either brlA or stuA blocked conidiation and altered hyphal morphology during growth on solid media, as shown by light and scanning electron microscopy, but did not affect biomass production during liquid-submerged growth. Genome-wide transcriptional profiling identified a complex StuA- and BrlA-dependent regulatory network, including genes previously shown to be involved in development and secondary metabolism. Remarkably, inactivation of stuA, but not brlA, drastically downregulated expression of the penicillin biosynthetic gene cluster during solid and liquid-submerged growth. In agreement, penicillin V production was wild-type-like in brlA-deficient strains but 99% decreased in stuA-deficient strains during liquid-submerged growth, as shown by high-performance liquid chromatography (HPLC) analysis. Thus, among identified regulators of penicillin V production StuA has the most severe influence. Overexpression of stuA increased the transcript levels of brlA and abaA (another developmental regulator) and derepressed conidiation during liquid-submerged growth but did not affect penicillin V productivity. Taken together, these data demonstrate an intimate but not exclusive link between regulation of development and secondary metabolism in P. chrysogenum.
Product Description
Roche
DIG Luminescent Detection Kit, sufficient for 50 blots (10 cm x 10 cm each), kit of 1 (5 components), suitable for hybridization
Sigma-Aldrich
REDExtract-N-Amp PCR ReadyMix, Ready-to-use 2X PCR Master Mix with Loading Dye
Roche
PCR DIG Probe Synthesis Kit, sufficient for 25 reaction (50 μL final reaction volume)