MilliporeSigma
Dissemination of 6S RNA among bacteria.
RNA biology (2014-12-09)
Stefanie Wehner, Katrin Damm, Roland K Hartmann, Manja Marz
ABSTRACT
6S RNA is a highly abundant small non-coding RNA widely spread among diverse bacterial groups. By competing with DNA promoters for binding to RNA polymerase (RNAP), the RNA regulates transcription on a global scale. RNAP produces small product RNAs derived from 6S RNA as template, which rearranges the 6S RNA structure leading to dissociation of 6S RNA:RNAP complexes. Although 6S RNA has been experimentally analysed in detail for some species, such as Escherichia coli and Bacillus subtilis, and was computationally predicted in many diverse bacteria, a complete and up-to-date overview of the distribution among all bacteria is missing. In this study we searched with new methods for 6S RNA genes in all currently available bacterial genomes. We ended up with a set of 1,750 6S RNA genes, of which 1,367 are novel and bona fide, distributed among 1,610 bacteria, and had a few tentative candidates among the remaining 510 assembled bacterial genomes accessible. We were able to confirm two tentative candidates by Northern blot analysis. We extended 6S RNA genes of the Flavobacteriia significantly in length compared to the present Rfam entry. We describe multiple homologs of 6S RNAs (including split 6S RNA genes) and performed a detailed synteny analysis.
Product Description
Roche
Nylon Membranes, positively charged, roll W × L 0.3 m × 3 m, sheet W × L 10 cm × 15 cm, sheet W × L 20 cm × 30 cm
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
DIG Wash and Block Buffer Set, storage temp.:2-8°C
Roche
DIG Easy Hyb Granules, powder, suitable for hybridization