MilliporeSigma
  • Transcriptional control of stem cell maintenance in the Drosophila intestine.
Transcriptional control of stem cell maintenance in the Drosophila intestine.
Development (Cambridge, England) (2010-02-12)
Allison J Bardin, Carolina N Perdigoto, Tony D Southall, Andrea H Brand, François Schweisguth
ABSTRACT
Adult stem cells maintain tissue homeostasis by controlling the proper balance of stem cell self-renewal and differentiation. The adult midgut of Drosophila contains multipotent intestinal stem cells (ISCs) that self-renew and produce differentiated progeny. Control of ISC identity and maintenance is poorly understood. Here we find that transcriptional repression of Notch target genes by a Hairless-Suppressor of Hairless complex is required for ISC maintenance, and identify genes of the Enhancer of split complex [E(spl)-C] as the major targets of this repression. In addition, we find that the bHLH transcription factor Daughterless is essential to maintain ISC identity and that bHLH binding sites promote ISC-specific enhancer activity. We propose that Daughterless-dependent bHLH activity is important for the ISC fate and that E(spl)-C factors inhibit this activity to promote differentiation.
Product Description
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep