MilliporeSigma
Single-molecule analysis of DNA replication in Xenopus egg extracts.
Methods (San Diego, Calif.) (2012-04-17)
Hasan Yardimci, Anna B Loveland, Antoine M van Oijen, Johannes C Walter
ABSTRACT
The recent advent in single-molecule imaging and manipulation methods has made a significant impact on the understanding of molecular mechanisms underlying many essential cellular processes. Single-molecule techniques such as electron microscopy and DNA fiber assays have been employed to study the duplication of genome in eukaryotes. Here, we describe a single-molecule assay that allows replication of DNA attached to the functionalized surface of a microfluidic flow cell in a soluble Xenopus leavis egg extract replication system and subsequent visualization of replication products via fluorescence microscopy. We also explain a method for detection of replication proteins, through fluorescently labeled antibodies, on partially replicated DNA immobilized at both ends to the surface.
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥13 units/mg protein
Roche
Anti-Digoxigenin-Fluorescein, Fab fragments, from sheep
Roche
Digoxigenin-11-dUTP, alkali-stable