MilliporeSigma
  • Systematic discovery of novel ciliary genes through functional genomics in the zebrafish.
Systematic discovery of novel ciliary genes through functional genomics in the zebrafish.
Development (Cambridge, England) (2014-08-21)
Semil P Choksi, Deepak Babu, Doreen Lau, Xianwen Yu, Sudipto Roy
ABSTRACT
Cilia are microtubule-based hair-like organelles that play many important roles in development and physiology, and are implicated in a rapidly expanding spectrum of human diseases, collectively termed ciliopathies. Primary ciliary dyskinesia (PCD), one of the most prevalent of ciliopathies, arises from abnormalities in the differentiation or motility of the motile cilia. Despite their biomedical importance, a methodical functional screen for ciliary genes has not been carried out in any vertebrate at the organismal level. We sought to systematically discover novel motile cilia genes by identifying the genes induced by Foxj1, a winged-helix transcription factor that has an evolutionarily conserved role as the master regulator of motile cilia biogenesis. Unexpectedly, we find that the majority of the Foxj1-induced genes have not been associated with cilia before. To characterize these novel putative ciliary genes, we subjected 50 randomly selected candidates to a systematic functional phenotypic screen in zebrafish embryos. Remarkably, we find that over 60% are required for ciliary differentiation or function, whereas 30% of the proteins encoded by these genes localize to motile cilia. We also show that these genes regulate the proper differentiation and beating of motile cilia. This collection of Foxj1-induced genes will be invaluable for furthering our understanding of ciliary biology, and in the identification of new mutations underlying ciliary disorders in humans.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Roche
T7 RNA Polymerase, from Escherichia coli BL 21/pAR 1219
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep