MilliporeSigma
  • Overexpression of Numb suppresses growth, migration, and invasion of human clear cell renal cell carcinoma cells.
Overexpression of Numb suppresses growth, migration, and invasion of human clear cell renal cell carcinoma cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-12-07)
Jin Sima, Bao Zhang, Yuanzi Yu, Xinyuan Sima, Yanxin Mao
ABSTRACT
The objective of the study was to investigate the impact of Numb on cell growth, cell migration, and invasion in human clear cell renal cell carcinoma (ccRCC). Endogenous expression of Numb was evaluated in the ccRCC cell lines (786-O, Caki-1, and Caki-2) and control reference human renal proximal tubular epithelial cells. Numb expression was decreased in the ccRCC cells compared with the control cells (P < 0.01). Then, 786-O and Caki-1 cells described as suitable transfection hosts were used in transfection to carry out biological function studies. The three experimental groups were as follows: Numb-ORF (transfected with Numb-ORF plasmid), blank-vector (transfected with pCMV6-entry), and cell-alone group (no DNA). Numb expression in the Numb-ORF groups was significantly higher than that in the controls (P < 0.01). Cell growth was remarkably reduced (P < 0.01), and the number of migrating or invading cells was reduced (P < 0.01) in the Numb-ORF groups compared with controls. Furthermore, the ratio of G0/G1 phase in the Numb-ORF group of 786-O cells was increased, and the S phase fraction and proliferation index was decreased (P < 0.01). Cyclin D1 and MMP-9 expression was reduced in the Numb-ORF groups compared with controls. Here, we have provided data for attenuated Numb expression in the ccRCC cells. Overexpression of Numb could induce G0/G1 phase arrest and inhibit cell proliferation, migration, and invasion. The suppressive effects might be due to downregulation of cyclin D1 or MMP-9 expression. Taken together, our data suggest that Numb may possibly function as a tumor suppressor involved in the carcinogenesis of ccRCC.