MilliporeSigma
6-Oxo and 6-thio purine analogs as antimycobacterial agents.
Bioorganic & medicinal chemistry (2013-02-26)
Ashish K Pathak, Vibha Pathak, Lainne E Seitz, William J Suling, Robert C Reynolds
ABSTRACT
6-Oxo and 6-thio analogs of purine were prepared based on the initial activity screening of a small, diverse purine library against Mycobacterium tuberculosis (Mtb). Certain 6-oxo and 6-thio-substituted purine analogs described herein showed moderate to good inhibitory activity. N(9)-substitution apparently enhances the anti-mycobacterial activity in the purine series described herein. Several 2-amino and 2-chloro purine analogs were also synthesized that showed moderate inhibitory activity against Mtb.