MilliporeSigma
  • Effect of juvenile hormone and serotonin (5-HT) on mixis induction of the rotifer Brachionus plicatilis Muller.
Effect of juvenile hormone and serotonin (5-HT) on mixis induction of the rotifer Brachionus plicatilis Muller.
Journal of experimental marine biology and ecology (2000-08-30)
Gallardo, Hagiwara, Snell
ABSTRACT
Juvenile hormone (JH) and serotonin (5-HT) were previously shown to enhance mictic (sexual) female production of the rotifer Brachionus plicatilis in batch cultures. To explore the basis of these effects, experiments were conducted on isolated individuals. JH treatment of maternal rotifers with 5 and 50 µgml(-1) (18.8 and 187.7 µM) resulted in significantly higher (P<0.05) mictic female production in the second (F(2)) and third (F(3)) generations. JH treatment was effective even at a lower food concentration of 7x10(5) cellsml(-1), but it was not effective when free ammonia was added at 2.4 and 3.1 µgml(-1). Mictic female production was not increased with exposure to 5-HT up to 50 µgml(-1) (129.1 µM) concentrations. When food level was reduced to 7x10(5) cellsml(-1), however, 5-HT-treated rotifers produced significantly (P<0.05) more mictic females than the control, particularly in F(3) generation. Mictic female production of 5-HT-treated rotifers did not differ from that of the control with or without free ammonia, but the intrinsic rate of natural increase (r) of 5-HT-treated rotifers at 3.1 µgml(-1) free ammonia was significantly higher than the control. These results show that juvenile hormone increases mictic female production under optimum and sub-optimum food levels, whereas 5-HT increases both mictic female production at low food level and population growth rate at high free ammonia concentrations. These compounds could be used to manage rotifer cultures and probe the mechanisms controlling the rotifer life cycle as it switches to mictic reproduction.