MilliporeSigma
Removal of colloidal biogenic selenium from wastewater.
Chemosphere (2015-01-07)
Lucian C Staicu, Eric D van Hullebusch, Mehmet A Oturan, Christopher J Ackerson, Piet N L Lens
ABSTRACT
Biogenic selenium, Se(0), has colloidal properties and thus poses solid-liquid separation problems, such as poor settling and membrane fouling. The separation of Se(0) from the bulk liquid was assessed by centrifugation, filtration, and coagulation-flocculation. Se(0) particles produced by an anaerobic granular sludge are normally distributed, ranging from 50 nm to 250 nm, with an average size of 166±29 nm and a polydispersity index of 0.18. Due to its nanosize range and protein coating-associated negative zeta potential (-15 mV to -23 mV) between pH 2 and 12, biogenic Se(0) exhibits colloidal properties, hampering its removal from suspension. Centrifugation at different centrifugal speeds achieved 22±3% (1500 rpm), 73±2% (3000 rpm) and 91±2% (4500 rpm) removal. Separation by filtration through 0.45 μm filters resulted in 87±1% Se(0) removal. Ferric chloride and aluminum sulfate were used as coagulants in coagulation-flocculation experiments. Aluminum sulfate achieved the highest turbidity removal (92±2%) at a dose of 10(-3) M, whereas ferric chloride achieved a maximum turbidity removal efficiency of only 43±4% at 2.7×10(-4) M. Charge repression plays a minor role in particle neutralization. The sediment volume resulting from Al2(SO3)4 treatment is three times larger than that produced by FeCl3.
Sigma-Aldrich
Aluminum sulfate octadecahydrate, ≥97%