MilliporeSigma
  • Postsynaptic density protein PSD-95 expression in Alzheimer's disease and okadaic acid induced neuritic retraction.
Postsynaptic density protein PSD-95 expression in Alzheimer's disease and okadaic acid induced neuritic retraction.
Neurobiology of disease (2008-04-22)
Geneviève Leuba, Claude Walzer, André Vernay, Béatrice Carnal, Rudolf Kraftsik, Françoise Piotton, Pascale Marin, Constantin Bouras, Armand Savioz
ABSTRACT
In order to understand how plasticity is related to neurodegeneration, we studied synaptic proteins with quantitative immunohistochemistry in the entorhinal cortex from Alzheimer patients and age-matched controls. We observed a significant decrease in presynaptic synaptophysin and an increase in postsynaptic density protein PSD-95, positively correlated with beta amyloid and phosphorylated Tau proteins in Alzheimer cases. Furthermore, Alzheimer-like neuritic retraction was generated in okadaic acid (OA) treated SH-SY5Y neuroblastoma cells with no decrease in PSD-95 expression. However, in a SH-SY5Y clone with decreased expression of transcription regulator LMO4 (as observed in Alzheimer's disease) and increased neuritic length, PSD-95 expression was enhanced but did not change with OA treatment. Therefore, increased PSD-95 immunoreactivity in the entorhinal cortex might result from compensatory mechanisms, as in the SH-SY5Y clone, whereas increased Alzheimer-like Tau phosphorylation is not related to PSD-95 expression, as suggested by the OA-treated cell models.