MilliporeSigma
  • A polymorphism in the promoter region of the selenoprotein S gene (SEPS1) contributes to Hashimoto's thyroiditis susceptibility.
A polymorphism in the promoter region of the selenoprotein S gene (SEPS1) contributes to Hashimoto's thyroiditis susceptibility.
The Journal of clinical endocrinology and metabolism (2014-01-30)
Liliana R Santos, Cecília Durães, Adélia Mendes, Hugo Prazeres, Maria Inês Alvelos, Carla Susete Moreira, Paulo Canedo, César Esteves, Celestino Neves, Davide Carvalho, Manuel Sobrinho-Simões, Paula Soares
ABSTRACT
The association between selenium and inflammation and the relevance of selenoproteins in follicular thyroid cell physiology have pointed to a putative role of selenoproteins in the pathogenesis of autoimmune thyroid diseases. The aim of this study was to evaluate the role of a promoter variation in SEPS1, the selenoprotein S gene, in the risk for developing Hashimoto's thyroiditis (HT). A case-control study was performed to assess the association of genetic variation in the SEPS1 gene (SEPS1 -105G/A single-nucleotide polymorphism, rs28665122) and HT. The study was conducted in north Portugal, Porto, in the period of 2007-2013. A total of 997 individuals comprising 481 HT patients and 516 unrelated controls were enrolled in the study. Genetic variants were discriminated by real-time PCR using TaqMan single-nucleotide polymorphism genotyping assays. There is a significant association between the SEPS1 -105 GA and AA genotypes and HT [odds ratio (OR) 2.24, confidence interval (CI) 1.67-3.02, P < 5.0 × 10(-7), and OR 2.08, CI 1.09-3.97, P = .0268, respectively]. The A allele carriers are in higher proportion in the patient group than in the control population (46.2% vs 28.1%, P < 5.0 × 10(-7)) with an OR (CI) of 2.22 (1.67-2.97). The proportion of patients carrying the A allele is significantly higher in male patients with HT, representing a 3.94 times increased risk (P = 7.9 × 10(-3)). Our findings support the existence of a link between SEPS1 promoter genetic variation and HT risk.