MilliporeSigma
  • MiR-197 induces Taxol resistance in human ovarian cancer cells by regulating NLK.
MiR-197 induces Taxol resistance in human ovarian cancer cells by regulating NLK.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-04-03)
Dongling Zou, Dong Wang, Rong Li, Ying Tang, Li Yuan, Xingtao Long, Qi Zhou
ABSTRACT
Chemotherapy is the preferred therapeutic approach for the therapy of advanced ovarian cancer, but 5-year survival rate remains low due to the development of drug resistance. Increasing evidence has documented that microRNAs (miRNAs) act important roles in drug resistance in a variety types of cancer. However, the roles of miRNA in regulating Taxol resistance in ovarian cancer and the detailed mechanism are less reported. We used Taqman probe stem loop real-time PCR to accurately measure the levels of miR-197 in normal ovarian cells, ovarian cancer cells, and Taxol-resistant ovarian cancer cells and found that miR-197 was significantly increased in Taxol-resistant ovarian cancer cells. Enforced expression of miR-197 can promote Taxol resistance, cell proliferation, and invasion of ovarian cancer cells. Meanwhile, repression of miR-197 in ovarian cancer cells can sensitize its response to Taxol and also induced attenuated cell proliferation and invasion ability. Furthermore, investigation of the detailed mechanism showed that the promotion of miR-197 on drug resistance in ovarian cancer cells was partially mediated by downregulating NLK, a negative regulator of WNT signaling pathway. Taken together, our work first demonstrated that miR-197 can confer drug resistance to Taxol, by regulating tumor suppressor, NLK expression in ovarian cancer cells.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nlk
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting human GPI
Sigma-Aldrich
MISSION® esiRNA, targeting human NLK