MilliporeSigma
  • Insulin receptor isoform A confers a higher proliferative capability to pancreatic beta cells enabling glucose availability and IGF-I signaling.
Insulin receptor isoform A confers a higher proliferative capability to pancreatic beta cells enabling glucose availability and IGF-I signaling.
Molecular and cellular endocrinology (2015-03-24)
Oscar Escribano, Almudena Gómez-Hernández, Sabela Díaz-Castroverde, Carmen Nevado, Gema García, Yolanda F Otero, Liliana Perdomo, Nuria Beneit, Manuel Benito
ABSTRACT
The main compensatory response to insulin resistance is the pancreatic beta cell hyperplasia to account for increased insulin secretion. In fact, in a previous work we proposed a liver-pancreas endocrine axis with IGF-I (insulin-like growth factor type I) secreted by the liver acting on IRA insulin receptor in beta cells from iLIRKO mice (inducible Liver Insulin Receptor KnockOut) that showed a high IRA/IRB ratio. However, the role of insulin receptor isoforms in the IGF-I-induced beta cell proliferation as well as the underlying molecular mechanisms remain poorly understood. For this purpose, we have used four immortalized mouse beta cell lines: bearing IR (IRLoxP), lacking IR (IRKO), expressing exclusively IRA (IRA), or alternatively expressing IRB (IRB). Pancreatic beta cell proliferation studies showed that IRA cells are more sensitive than those expressing IRB to the mitogenic response induced by IGF-I, acting through the pathway IRA/IRS-1/2/αp85/Akt/mTORC1/p70S6K. More importantly, IRA beta cells, but not IRB, showed an increased glucose uptake as compared with IRLoxP cells, this effect being likely owing to an enhanced association between Glut-1 and Glut-2 with IRA. Overall, our results strongly suggest a prevalent role of IRA in glucose availability and IGF-I-induced beta cell proliferation mainly through mTORC1. These results could explain, at least partially, the role played by the liver-secreted IGF-I in the compensatory beta cell hyperplasia observed in response to severe hepatic insulin resistance in iLIRKO mice.
Sigma-Aldrich
MISSION® esiRNA, targeting human INSR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Insr