Sigma-Aldrich
Calcium chloride, AnhydroBeads™, −10 mesh, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium chloride, AnhydroBeads™, −10 mesh, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium chloride
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max™, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Vetec™, reagent grade, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M