MilliporeSigma
  • MicroRNA-204 suppresses trophoblast-like cell invasion by targeting matrix metalloproteinase-9.
MicroRNA-204 suppresses trophoblast-like cell invasion by targeting matrix metalloproteinase-9.
Biochemical and biophysical research communications (2015-05-25)
Yang Yu, Leilei Wang, Tong Liu, Hongbo Guan
ABSTRACT
Preeclampsia is a devastating pregnancy-related syndrome characterized by the onset of hypertension, proteinuria and edema. Insufficient invasion of trophoblasts is well-known to be correlated with preeclampsia development. The present study was performed to investigate the functional role microRNA (miRNA)-204 in trophoblastic invasion in vitro. We here found that the invasive capabilities of BeWo and JEG3 trophoblast-like cells were suppressed by miR-204 mimics, whereas enhanced by its inhibitor. Matrix metalloproteinase-9 (MMP9) was first confirmed to play a role in regulating trophoblast invasion through loss- or gain-of-function experiment. Notably, we demonstrated MMP9 as a direct target of miR-204 in BeWo cells by using the dual-luciferase assay. Moreover, forced overexpression of MMP9 was noted to partly attenuate the inhibitory effects of miR-204 on BeWo cell invasion. Taken together, our study indicates that miR-204 may contribute to the development of preeclampsia by inhibiting trophoblastic invasion, and that MMP9 is involved in miR-204-mediated trophoblast cell invasion. Our study suggests miR-204 as a novel therapeutic target for preeclampsia.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP9
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mmp9