MilliporeSigma
  • Identification and Characterization of CD300H, a New Member of the Human CD300 Immunoreceptor Family.
Identification and Characterization of CD300H, a New Member of the Human CD300 Immunoreceptor Family.
The Journal of biological chemistry (2015-07-30)
Kouta Niizuma, Satoko Tahara-Hanaoka, Emiko Noguchi, Akira Shibuya
ABSTRACT
Recruitment of circulating monocytes and neutrophils to infection sites is essential for host defense against infections. Here, we identified a previously unannotated gene that encodes an immunoglobulin-like receptor, designated CD300H, which is located in the CD300 gene cluster. CD300H has a short cytoplasmic tail and associates with the signaling adaptor proteins, DAP12 and DAP10. CD300H is expressed on CD16(+) monocytes and myeloid dendritic cells. Ligation of CD300H on CD16(+) monocytes and myeloid dendritic cells with anti-CD300H monoclonal antibody induced the production of neutrophil chemoattractants. Interestingly, CD300H expression varied among healthy subjects, who could be classified into two groups according to "positive" and "negative" expression. Genomic sequence analysis revealed a single-nucleotide substitution (rs905709 (G → A)) at a splice donor site on intron 1 on either one or both alleles. The International HapMap Project database has demonstrated that homozygosity for the A allele of single nucleotide polymorphism (SNP) rs905709 ("negative" expression) is highly frequent in Han Chinese in Beijing, Japanese in Tokyo, and Europeans (A/A genotype frequencies 0.349, 0.167, and 0.138, respectively) but extremely rare in Sub-Saharan African populations. Together, these results suggest that CD300H may play an important role in innate immunity, at least in populations that carry the G/G or G/A genotype of CD300H.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
trans-2-Bromovinylboronic acid MIDA ester, technical grade
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Digitonin, ~50% (TLC)