MilliporeSigma
The determinants of efficiency in the Canadian health care system.
Health economics, policy, and law (2015-05-23)
Sara Allin, Michel Grignon, Li Wang
ABSTRACT
In spite of the vast number of studies measuring economic efficiency in health care, there has been little take-up of this evidence by policy-makers to date. This study provides an illustration of how a system-level study drawing on best practice in empirical measurement of efficiency may be of practical use to health system decision makers and managers. We make use of the rich data available in Canada to undertake a robust two-stage data envelopment analysis to calculate efficiency at the regional (sub-provincial) level. Decisions about what the health system produces (the outcome to measure efficiency against) and what are the resources it has to produce that outcome were based on interviews and consultation with health system decision makers. Overall, we find large inefficiencies in the Canadian health care system, which could improve outcomes (here, measured as a reduction in treatable causes of death) by between 18 and 35% across our analyses. Also, we find that inefficiencies are the result of three main sets of factors that policy makers could pay attention to: management factors, such as hospital re-admissions; public health factors, such as obesity and smoking rates; and environmental factors such as the population's average income.
Sigma-Aldrich
N,N′-Bis(hydroxymethyl)urea, ≥95%
Sigma-Aldrich
N,N-Diethylaniline, ≥99%