MilliporeSigma
  • Necrosis-induced TLR3 Activation Promotes TLR2 Expression in Gingival Cells.
Necrosis-induced TLR3 Activation Promotes TLR2 Expression in Gingival Cells.
Journal of dental research (2015-06-06)
K Mori, M Yanagita, S Hasegawa, M Kubota, M Yamashita, S Yamada, M Kitamura, S Murakami
ABSTRACT
Damage-associated molecular patterns (DAMPs), endogenous molecules released from injured or dying cells, evoke sterile inflammation that is not induced by microbial pathogens. Periodontal diseases are infectious diseases caused by oral microorganisms; however, in some circumstances, DAMPs might initiate inflammatory responses before host cells recognize pathogen-associated molecular patterns. Here, we showed that the necrotic cell supernatant (NCS) functioned as an endogenous danger signal when released from necrotic epithelial cells exposed to repeat freeze thawing. The NCS contained RNA and stimulated the production of inflammatory cytokines interleukin 6 (IL-6) and IL-8 from gingival epithelial cells and gingival fibroblasts. Targeted knockdown of Toll-like receptor 3 (TLR3) in these cells significantly suppressed the ability of the NCS to induce IL-6 and IL-8 production. Epithelial cells and fibroblasts recognized the NCS from heterologous cells. Interestingly, the activation of TLR3, rather than other TLRs, induced TLR2 mRNA expression and proteins in gingival epithelial cells, and pretreatment with the NCS or polyinosinic:polycytidylic acid (Poly(I:C)), a strong TLR3 activator, enhanced inflammatory cytokine production induced by subsequent stimulation with Porphyromonas gingivalis (P. gingivalis) lipopolysaccharide, a TLR2 agonist. Moreover, the NCS reduced the expression of epithelial tight junction molecules zona occludens 1 and occludin and increased the permeability of epithelial tight junctions. These findings suggest that endogenous danger signal molecules such as self-RNA released from necrotic cells are recognized by TLR3 and that a subsequent increase of TLR2 expression in periodontal compartments such as gingival epithelial cells and gingival fibroblasts may enhance the inflammatory response to periodontopathic microbes recognized by TLR2 such as P. gingivalis, which also disrupts epithelial barrier functions. Thus, DAMPs may be involved in the development and prolongation of periodontal disease.
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR3
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tlr3