MilliporeSigma
  • Live-cell imaging of actin dynamics reveals mechanisms of stereocilia length regulation in the inner ear.
Live-cell imaging of actin dynamics reveals mechanisms of stereocilia length regulation in the inner ear.
Nature communications (2015-04-22)
Meghan C Drummond, Melanie Barzik, Jonathan E Bird, Duan-Sun Zhang, Claude P Lechene, David P Corey, Lisa L Cunningham, Thomas B Friedman
ABSTRACT
The maintenance of sensory hair cell stereocilia is critical for lifelong hearing; however, mechanisms of structural homeostasis remain poorly understood. Conflicting models propose that stereocilia F-actin cores are either continually renewed every 24-48 h via a treadmill or are stable, exceptionally long-lived structures. Here to distinguish between these models, we perform an unbiased survey of stereocilia actin dynamics in more than 500 utricle hair cells. Live-imaging EGFP-β-actin or dendra2-β-actin reveal stable F-actin cores with turnover and elongation restricted to stereocilia tips. Fixed-cell microscopy of wild-type and mutant β-actin demonstrates that incorporation of actin monomers into filaments is required for localization to stereocilia tips. Multi-isotope imaging mass spectrometry and live imaging of single differentiating hair cells capture stereociliogenesis and explain uniform incorporation of (15)N-labelled protein and EGFP-β-actin into nascent stereocilia. Collectively, our analyses support a model in which stereocilia actin cores are stable structures that incorporate new F-actin only at the distal tips.
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%