MilliporeSigma
  • A new effective on chip electromembrane extraction coupled with high performance liquid chromatography for enhancement of extraction efficiency.
A new effective on chip electromembrane extraction coupled with high performance liquid chromatography for enhancement of extraction efficiency.
Analytica chimica acta (2015-11-04)
Yousef Abdossalami Asl, Yadollah Yamini, Shahram Seidi, Behnam Ebrahimpour
ABSTRACT
In the present research, an effective on chip electromembrane extraction (CEME) coupled with high performance liquid chromatography was presented for analysis of nortriptyline (NOR) and amitriptyline (AMI) as basic model analytes from urine samples. The chip consists of two polymethyl methacrylate (PMMA) parts with two craved microfluidic channels in each part. These channels were used as flow path for the sample solution and a thin compartment for the acceptor phase. A porous polypropylene sheet membrane impregnated with an organic solvent was placed between two parts of chip device to separate the channels. Two platinum electrodes were mounted at the bottom of these channels that were connected to a power supply providing the electrical driving force for migration of ionized analytes from sample solution through the porous sheet membrane into the acceptor phase. This new setup provides effective and reproducible extractions with low volume of sample solution. Efficient parameters on CEME of the model analytes were optimized using one variable at a time method. Under the optimized conditions, the calibration curve was linear in the range of 10.0-500 μg L(-1) with coefficient of determination (r(2)) more than 0.9902. The relative standard deviations (RSDs %) for extraction and determination of the analytes were less than 6.8% based on six replicate measurements. LODs less than 4.0 μg L(-1) were obtained for both of the model analytes. The preconcentration factors higher than 17.0-fold were obtained. The results demonstrated that CEME would be used efficiently for extraction and determination of AMI and NOR from urine samples.
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Undecanol, 97%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Tris(2-ethylhexyl) phosphate, 97%
Sigma-Aldrich
2-Nitrophenyl octyl ether, 99%
Sigma-Aldrich
Undecyl alcohol, 97%, FG