MilliporeSigma
  • Composite films of polydopamine-Alcian Blue for colored coating with new physical properties.
Composite films of polydopamine-Alcian Blue for colored coating with new physical properties.
Journal of colloid and interface science (2015-08-12)
Florian Ponzio, Jérôme Bour, Vincent Ball
ABSTRACT
Polydopamine (PDA) coatings appear as a universal functionalization methodology allowing to coat the surface of almost all kinds of known materials with a conformal, stable, robust and reactive material. Relatively few investigations were dedicated to the incorporation of other molecules in PDA coatings during their deposition from dopamine solutions under oxidative conditions. Herein we rely on the assumption that the basic building blocks of PDA could be porphyrin like tetramers (as well as higher order oligomers) of 5,6-dihydroxyindole and we investigate the influence of a cationic Cu(II) phtalocyanine, namely Alcian Blue (AB), on the deposition kinetics and on the properties of PDA films. We demonstrate that AB is indeed incorporated in the PDA films to yield a composite PDA-AB coating displaying the optical features of both PDA and AB. The amount of incorporated dye depends on its concentration in solution. The obtained PDA-AB films have a smaller thickness than their related PDA counterparts, a different morphology and a higher permeability to the anionic hexacyanoferrate redox probe. In addition, the incorporation of AB in the films is not homogeneous through their thickness as inferred by means of X-ray photoelectron spectroscopy. The reason for this interesting finding is discussed on the basis of the interactions between AB and PDA as well as on the basis of the structure of PDA films.
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Supelco
Dopamine hydrochloride solution, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®