MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Deciphering the signaling mechanisms of the plant cell wall degradation machinery in Aspergillus oryzae.
Deciphering the signaling mechanisms of the plant cell wall degradation machinery in Aspergillus oryzae.
BMC systems biology (2015-11-18)
D B R K Gupta Udatha, Evangelos Topakas, Margarita Salazar, Lisbeth Olsson, Mikael R Andersen, Gianni Panagiotou
ABSTRACT
The gene expression and secretion of fungal lignocellulolytic enzymes are tightly controlled at the transcription level using independent mechanisms to respond to distinct inducers from plant biomass. An advanced systems-level understanding of transcriptional regulatory networks is required to rationally engineer filamentous fungi for more efficient bioconversion of different types of biomass. In this study we focused on ten chemically defined inducers to drive expression of cellulases, hemicellulases and accessory enzymes in the model filamentous fungus Aspergillus oryzae and shed light on the complex network of transcriptional activators required. The chemical diversity analysis of the inducers, based on 186 chemical descriptors calculated from the structure, resulted into three clusters, however, the global, metabolic and extracellular protein transcription of the A. oryzae genome were only partially explained by the chemical similarity of the enzyme inducers. Genes encoding enzymes that have attracted considerable interest such as cellobiose dehydrogenases and copper-dependent polysaccharide mono-oxygenases presented a substrate-specific induction. Several homology-model structures were derived using ab-initio multiple threading alignment in our effort to elucidate the interplay of transcription factors involved in regulating plant-deconstructing enzymes and metabolites. Systematic investigation of metabolite-protein interactions, using the 814 unique reactants involved in 2360 reactions in the genome scale metabolic network of A. oryzae, was performed through a two-step molecular docking against the binding pockets of the transcription factors AoXlnR and AoAmyR. A total of six metabolites viz., sulfite (H2SO3), sulfate (SLF), uroporphyrinogen III (UPGIII), ethanolamine phosphate (PETHM), D-glyceraldehyde 3-phosphate (T3P1) and taurine (TAUR) were found as strong binders, whereas the genes involved in the metabolic reactions that these metabolites appear were found to be significantly differentially expressed when comparing the inducers with glucose. Based on our observations, we believe that specific binding of sulfite to the regulator of the cellulase gene expression, AoXlnR, may be the molecular basis for the connection of sulfur metabolism and cellulase gene expression in filamentous fungi. Further characterization and manipulation of the regulatory network components identified in this study, will enable rational engineering of industrial strains for improved production of the sophisticated set of enzymes necessary to break-down chemically divergent plant biomass.
Sigma-Aldrich
Sophorose, ≥98% (HPLC, anhydrous)