MilliporeSigma
PAK1-mediated MORC2 phosphorylation promotes gastric tumorigenesis.
Oncotarget (2015-04-19)
Guiling Wang, Yanyan Song, Tong Liu, Chunyu Wang, Qing Zhang, Furong Liu, Xinze Cai, Zhifeng Miao, Hongde Xu, Huimian Xu, Liu Cao, Feng Li
ABSTRACT
To date, microrchidia (MORC) family CW-type zinc-finger 2 (MORC2), has been found to be involved in p21-activated kinase1 (PAK1) pathway to maintain genomic integrity. Here, we explore its novel role in cancer. We demonstrate that PAK1-mediated MORC2 phosphorylation promotes cell cycle progression, defective phosphorylation of MORC2-S677A results in attenuated cell proliferation and tumorigenicity of gastric cancer cells, which is significantly enhanced in overexpression of phospho-mimic MORC2-S677E form, suggesting the importance of MORC2 phosphorylation in tumorigenesis. More importantly, phosphorylation of MORC2 correlates positively with PAK1 expression in clinical gastric cancer. Furthermore, high expression of PAK1 and phosphorylation of MORC2 appear to be associated with poor prognosis of clinical gastric cancer. Collectively, these findings revealed a novel function of MORC2 phosphorylation in promoting gastric cell proliferation in vitro and tumorigenesis in vivo, suggesting that blocking PAK1-mediated MORC2 phosphorylation might be a potential therapeutic strategy for gastric tumorigenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pak1
Sigma-Aldrich
MISSION® esiRNA, targeting human PAK1
Sigma-Aldrich
MISSION® esiRNA, targeting human PKN1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stk3