MilliporeSigma
  • A Challenge Beyond Bottom Cells: Top-Illuminated Flexible Organic Solar Cells with Nanostructured Dielectric/Metal/Polymer (DMP) Films.
A Challenge Beyond Bottom Cells: Top-Illuminated Flexible Organic Solar Cells with Nanostructured Dielectric/Metal/Polymer (DMP) Films.
Advanced materials (Deerfield Beach, Fla.) (2015-06-04)
Juyoung Ham, Wan Jae Dong, Jae Yong Park, Chul Jong Yoo, Illhwan Lee, Jong-Lam Lee
ABSTRACT
Top-illuminated flexible organic solar cells with a high power conversion efficiency (≈6.75%) are fabricated using a dielectric/metal/polymer (DMP) electrode. Employing a polymer layer (n = 1.49) makes it possible to show the high transmittance, which is insensitive to film thickness, and the excellent haze induced by well-ordered nanopatterns on the DMP electrode, leading to a 28% of enhancement in efficiency compared to bottom cells.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Indium tin oxide coated PET, surface resistivity 60 Ω/sq, L × W × thickness 1 ft × 1 ft × 5 mil, sheet
Tantalum(V) ethoxide, packaged for use in deposition systems