MilliporeSigma
  • Down-regulation of mir-542-3p promotes neointimal formation in the aging rat.
Down-regulation of mir-542-3p promotes neointimal formation in the aging rat.
Vascular pharmacology (2015-06-01)
De-hui Qian, Pan Gao, Huan Feng, Zhe-Xue Qin, Jia-bei Li, Lan Huang
ABSTRACT
To explore mir-542-3p mediated inhibition of vascular smooth muscle cell (VSMC) proliferation through the inhibition of Syk activation. MicroRNA (mir)-542-3p was selected for analysis based on miRNA microarray and qRT-PCR results. In vitro mir-542-3p expression was significantly downregulated in old (o)VSMCs compared with young (y)VSMCs under serum stimulation conditions. Upregulation of mir-542-3p in oVSMCs significantly inhibited VSMC proliferation, whereas downregulation of mir-542-3p in yVSMCs increased VSMC proliferation. We identified spleen tyrosine kinase (Syk) as a direct target of mir-542-3p by database search, and showed that its expression and phosphorylation were higher in oVSMCs than in yVSMCs after serum stimulation. Luciferase assays confirmed that Syk is a direct target of miR-3542-3p. Knock-down of mir-542-3p in yVSMCs inhibited the activation of the Syk downstream effectors STAT3 and STAT5, whereas mir-542-3p overexpression enhanced STAT3 and STAT5 activities. In a rat balloon injury model, mir-542-3p inhibited neointima formation and proliferating cell nuclear antigen (PCNA) protein expression. Mir-542-3p modulates VSMC proliferation via the Syk/STAT3-STAT5 axis. Downregulation of mir-542-3p may explain age-related neointimal hyperplasia in rats.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
Sodium chloride solution, 5 M