MilliporeSigma
  • Serum IGFBP7 levels associate with insulin resistance and the risk of metabolic syndrome in a Chinese population.
Serum IGFBP7 levels associate with insulin resistance and the risk of metabolic syndrome in a Chinese population.
Scientific reports (2015-05-20)
Yi Liu, Minliang Wu, Jie Ling, Libing Cai, Dandan Zhang, Harvest F Gu, Hao Wang, Yimin Zhu, Maode Lai
ABSTRACT
Metabolic syndrome (MetS), one of the major public health concerns, is regarded as the "common soil" of incidence of common chronic diseases and may increase the risk of type 2 diabetes. The predominant underlying mechanism of MetS is insulin resistance (IR). Additionally, previous studies have indicated that IGFBP7 has high affinity of binding with insulin and might induce IR. The objective of this study was to firstly evaluate the associations of serum IGFBP7 levels with IR and MetS with a relatively large sample and population based design. In a population based MetS case-control study, HOMA-IR was used to evaluate the insulin sensitivity and serum IGFBP7 levels were determined with chemiluminescence-linked immunoassay. As a result, the subjects of MetS and IR had higher serum levels of IGFBP7 than control healthy subjects. High serum IGFBP7 levels increased the risk of MetS and IR. Serum IGFBP7 levels were also found to be significantly correlated with metabolic-associated parameters of Waist-to-hip ratio (WHR), HDL and LDL. These findings suggest that serum IGFBP7 levels are associated with IR and MetS, providing new insight into the mechanism of IR and Mets. IGFBP7 may be a potential interventional target for IR and Mets.
Sigma-Aldrich
IGF-Binding Protein 7 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
SILuLite IGFBP7 Insulin-like growth factor-binding protein 7 human, recombinant, expressed in HEK 293 cells, MS Protein Standard