MilliporeSigma
  • Antimicrobial activity, DNA cleavage, thermal analysis data and crystal structure of some new CdLX2 complexes: A supramolecular network.
Antimicrobial activity, DNA cleavage, thermal analysis data and crystal structure of some new CdLX2 complexes: A supramolecular network.
Materials science & engineering. C, Materials for biological applications (2015-06-29)
M Montazerozohori, A Nazaripour, A Masoudiasl, R Naghiha, M Dusek, M Kucerakova
ABSTRACT
Some new cadmium(II) Schiff base complexes CdLX2 (where X=Cl(-), Br(-), I(-), SCN(-) and N3(-) and L=N,N-bis((E)-3-(4-(dimethylamino)phenyl)allylidene)benzene-1,2-diamine) were synthesized and characterized by elemental analysis, FT-IR, UV-visible, (1)HNMR spectra, and conductivity measurements. Moreover, the structure of the CdLI2 was determined by single crystal X-ray crystallography. CdLI2 crystallizes in a triclinic system with space group P 1̅. The molecular structure shows distorted tetrahedral coordination geometry. The CH....π and π-π stacking interactions connect the molecules to a supramolecular network. The thermal properties of the compounds were investigated from the room temperature to 800 °C with a heating rate of 10 °C/min and decomposition activation parameters were evaluated from the TG/DTG plots. Antibacterial/antifungal activities of the compounds were screened by the disk diffusion method against the Gram-negative bacteria Escherichia coli and Pseudomonas aeruginosa, the Gram-positive bacteria Staphylococcus aureus and Bacillus subtilis and the fungi strain Aspergillus niger and Candida albicans. The antimicrobial activities were determined for all compounds and the complexation was found to enhance the inhibitory activity. Finally, the DNA cleavage potential of the compounds was investigated by agarose gel electrophoresis method.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
4-(Dimethylamino)cinnamaldehyde, chromogenic reagent for indoles and flavanols
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%