MilliporeSigma
  • Effects of dietary trans-9 octadecenoic acid, trans-11 vaccenic acid and cis-9, trans-11 conjugated linoleic acid in mice.
Effects of dietary trans-9 octadecenoic acid, trans-11 vaccenic acid and cis-9, trans-11 conjugated linoleic acid in mice.
Molecular medicine reports (2015-05-13)
Tao Wang, Ji-Na Lim, Jae-Sung Lee, Sang-Bum Lee, Jin-Hee Hwang, U-Suk Jung, Min-Jeong Kim, Dae-Youn Hwang, Sang-Rak Lee, Sang-Gun Roh, Hong-Gu Lee
ABSTRACT
The aim of the present study was to investigate the effects of dietary trans fatty acids in mice. Following the administration of a 0.5/100 g diet of trans-9 octadecenoic acid (EA), trans-11 vaccenic acid (TVA) or cis-9, trans-11 conjugated linoleic acid (CLA) for 4 weeks, the body weights and the weights of the liver, testis and mediastinal adipose tissue (MAT) of the animals gradually decreased (P<0.05). The EA group exhibited the lowest levels of magnesium and triglycerides (P<0.05). CLA increased villus length (P<0.05), while EA and TVA decreased villus length (P<0.05). The TVA group exhibited the lowest levels of low-density lipoprotein and tumor necrosis factor-α (P<0.05). Taken together, EA, TVA and CLA affected the physiological conditions of mice differently. The potential effects of three well-known fatty acids, including trans-9 octadecenoic acid (EA), trans-11 vaccenic acid (TVA) and cis-9, trans-11 conjugated linoleic acid (CLA), in animals or humans remain to be elucidated. Therefore, in the present study, 32 animals were randomly divided into four groups and administered a 0.5/100 g diet of EA, TVA or CLA for 4 weeks. The results demonstrated that the body weights and the weights of the liver, testis and mediastinal adipose tissue (MAT) of the animals gradually decreased (P<0.05). Blood was collected individually via the external jugular veins and the EA group exhibited the lowest levels of magnesium and triglycerides (P<0.05). CLA increased villus length (P<0.05), while EA and TVA decreased villus length (P<0.05). The TVA group exhibited the lowest levels of low-density lipoprotein and tumor necrosis factor-α (P<0.05). Taken together, EA, TVA and CLA affected the physiological conditions of mice differently and these may further our understanding of the various effects of these fatty acids on animals and humans.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Boron trifluoride-methanol solution, 50% w/w in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Boron trifluoride methanol complex solution, 13-15% BF3 basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M