MilliporeSigma
  • New antimony(III) halide complexes with dithiocarbamate ligands derived from thiuram degradation: The effect of the molecule's close contacts on in vitro cytotoxic activity.
New antimony(III) halide complexes with dithiocarbamate ligands derived from thiuram degradation: The effect of the molecule's close contacts on in vitro cytotoxic activity.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
O S Urgut, I I Ozturk, C N Banti, N Kourkoumelis, M Manoli, A J Tasiopoulos, S K Hadjikakou
ABSTRACT
Antimony(III) halide complexes of the formulae {[SbBr(Me2DTC)2]n} (1), {[SbI(Me2DTC)2]n} (2) and {[(Me2DTC)2Sb(μ2-I)Sb(Me2DTC)2](+).I3(-)} (3) (Me2DTC = dimethyldithiocarbomate) were synthesized from SbX3, (X = Br or I) and tetramethylthiuram monosulfide (Me4tms) or tetramethylthiuram disulfide (Me4tds). The complexes were characterized by melting point (m.p.), elemental analysis (e.a.), Fourier-transform Infra-Red (FT-IR), Fourier-transform Raman (FT-Raman), Nuclear Magnetic Resonance ((1)H,(13)C-NMR) spectroscopy and Thermogravimetric-Differential Thermal Analysis (TG-DTA). Crystal structures of complexes 1-3 were determined with single crystal X-ray diffraction analysis. Complexes 1 and 2 are polymers with distorted square pyramidal (SP) geometry in each monomeric unit, whereas complex 3 is ionic, containing an iodonium linkage Sb-I(+)-Sb and an I3(-) counter anion; to the best of our knowledge, this is the first ionic antimony(III) iodide complex. The in vitro cytotoxic activity of 1-3 against human adenocarcinoma cells: breast (MCF-7) and cervix (HeLa) cells and non-cancerous cells: MRC-5 (normal human fetal lung fibroblast cells) was evaluated with trypan blue (TB) and sulforhodamine B (SRB) assays. Among antimony(III) compounds with sulfur containing ligand, those of dithiocarbamates exhibit significant cytotoxic activity. Hirshfeld surface volumes were analyzed to clarify the nature of the intermolecular interactions by the 2D fingerprint plot. Molecules with lower H-all atoms inter-molecular interactions exhibit the higher activity against MCF-7 cells. The in vivo genotoxicity of 1-3 was evaluated by the mean of Allium cepa test. Alterations in the mitotic index values due to the chromosomal aberrations were observed in the case of complexes 2 and 3. Since, no such alteration is caused by 1, it makes this compound candidate for further study as potential drug.
Sigma-Aldrich
Antimony(III) iodide, 98%
Sigma-Aldrich
Tetramethylthiuram monosulfide, 97%
Sigma-Aldrich
Antimony(III) chloride, ≥99.95% trace metals basis
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Antimony(III) chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Antimony(III) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Tetramethylthiuram disulfide, 97%
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade