MilliporeSigma
  • Fbxo25 controls Tbx5 and Nkx2-5 transcriptional activity to regulate cardiomyocyte development.
Fbxo25 controls Tbx5 and Nkx2-5 transcriptional activity to regulate cardiomyocyte development.
Biochimica et biophysica acta (2015-03-01)
Hoe-Su Jeong, Eun-Shil Jung, Ye-Ji Sim, Su-Jin Kim, Jae-Woo Jang, Ki-Sung Hong, Won-Young Lee, Hyung-Min Chung, Kyung-Tae Park, Yi-Sook Jung, Chang-Hoon Kim, Kye-Seong Kim
ABSTRACT
The ubiquitin-proteasome system (UPS) plays an important role in protein quality control, cellular signalings, and cell differentiation through the regulated turnover of key transcription factors in cardiac tissue. However, the molecular mechanism underlying Fbxo25-mediated ubiquitination of cardiac transcription factors remains elusive. We report that an Fbxo25-mediated SCF ubiquitination pathway regulates the protein levels and activities of Tbx5 and Nkx2-5 based on our studies using MG132, proteasome inhibitor, and the temperature sensitive ubiquitin system in ts20 cells. Our data indicate that Fbxo25 directly interacts with Tbx5 and Nkx2-5 in vitro and in vivo. In support of our findings, a dominant-negative mutant of Fbxo25, Fbxo251-236, prevents Tbx5 degradation and increases Tbx5 transcriptional activity in a Tbx5 responsive luciferase assay. Therefore, Fbxo25 facilitates Tbx5 degradation in an SCF-dependent manner. In addition, the silencing of endogenous Fbxo25 increases Tbx5 and Nkx2-5 mRNA levels and suppresses mESC-derived cardiomyocyte differentiation. Likewise, the exogenous expression of FBXO25 downregulates NKX2-5 level in human ESC-derived cardiomyocytes. In myocardial infarction model, Fbxo25 mRNA decreases, whereas the mRNA and protein levels of Tbx5 and Nkx2-5 increase. The protein levels of Tbx5 and Nkx2-5 are regulated negatively by Fbxo25-mediated SCF ubiquitination pathway. Thus, our findings reveal a novel mechanism for regulation of SCFFbox25-dependent Nkx2-5 and Tbx5 ubiquitination in cardiac development and provide a new insight into the regulatory mechanism of Nkx2-5 and Tbx5 transcriptional activity.
Sigma-Aldrich
MISSION® esiRNA, targeting human CACNA1C (2)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MISSION® esiRNA, targeting human CACNA1C (1)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cacna1c
Sigma-Aldrich
MISSION® esiRNA, targeting human TYMS
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tyms
Sigma-Aldrich
MISSION® esiRNA, targeting human TBXAS1
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)