MilliporeSigma
  • Mechanosensitive store-operated calcium entry regulates the formation of cell polarity.
Mechanosensitive store-operated calcium entry regulates the formation of cell polarity.
Journal of cellular physiology (2015-02-03)
Yi-Wei Huang, Shu-Jing Chang, Hans I-Chen Harn, Hui-Ting Huang, Hsi-Hui Lin, Meng-Ru Shen, Ming-Jer Tang, Wen-Tai Chiu
ABSTRACT
Ca(2+) -mediated formation of cell polarity is essential for directional migration which plays an important role in physiological and pathological processes in organisms. To examine the critical role of store-operated Ca(2+) entry, which is the major form of extracellular Ca(2+) influx in non-excitable cells, in the formation of cell polarity, we employed human bone osteosarcoma U2OS cells, which exhibit distinct morphological polarity during directional migration. Our analyses showed that Ca(2+) was concentrated at the rear end of cells and that extracellular Ca(2+) influx was important for cell polarization. Inhibition of store-operated Ca(2+) entry using specific inhibitors disrupted the formation of cell polarity in a dose-dependent manner. Moreover, the channelosomal components caveolin-1, TRPC1, and Orai1 were concentrated at the rear end of polarized cells. Knockdown of TRPC1 or a TRPC inhibitor, but not knockdown of Orai1, reduced cell polarization. Furthermore, disruption of lipid rafts or overexpression of caveolin-1 contributed to the downregulation of cell polarity. On the other hand, we also found that cell polarity, store-operated Ca(2+) entry activity, and cell stiffness were markedly decreased by low substrate rigidity, which may be caused by the disorganization of actin filaments and microtubules that occurs while regulating the activity of the mechanosensitive TRPC1 channel.
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)